Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$499.50

Specifications

Antigen DPP6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159427
SDP
View Documents
Novus Biologicals
NBP159427
100 μL
Each of 1 for $499.50
Only null left
Add to Cart
 
Description

Description

DPP6 Polyclonal specifically detects DPP6 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

DPP6
Polyclonal
Rabbit
P42658
1804
Synthetic peptides corresponding to DPP6(dipeptidyl-peptidase 6) The peptide sequence was selected from the middle region of DPP6. Peptide sequence FLIIHPTADEKIHFQHTAELITQLIRGKANYSLQIYPDESHYFTSSSLKQ.
Primary
Western Blot
Unconjugated
RUO
dipeptidyl aminopeptidase IV-related protein, dipeptidyl aminopeptidase-like protein 6, Dipeptidyl aminopeptidase-related protein, Dipeptidyl peptidase 6, Dipeptidyl peptidase IV-like protein, dipeptidyl peptidase IV-related protein, Dipeptidyl peptidase VI, dipeptidylpeptidase 6, dipeptidyl-peptidase 6, dipeptidylpeptidase VI, DPP VI, DPPXFLJ55680, MGC46605, VF2
DPP6
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.