Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562201 Supplier Invitrogen™ Supplier No. PA562201
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VKKAINDRQM PKVEYRDIEI DDYNLPMQIL KPATFTDTTH YPLLLVVDGT PGSQSVAEKF EVSWETVMVS SHGAVVVKCD G Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

DPP6 encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.60 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPP6
Gene Accession No. P42658, Q9Z218
Gene Alias B930011P16Rik; D5Buc3; D5Buc4; D5Buc5; dipeptidyl aminopeptidase IV-related protein; dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase 6; Dipeptidyl peptidase IV-like protein; dipeptidyl peptidase IV-related protein; dipeptidyl peptidase like 6; dipeptidyl peptidase VI; dipeptidylpeptidase 6; dipeptidyl-peptidase 6; dipeptidylpeptidase VI; DPL1; DPP VI; Dpp6; Dpp-6; DPPX; Gm1377; In(5)6H p; In(5)6H-p; inversion, Chr 5, Harwell 6, proximal; MRD33; Peplb; Rw; VF2
Gene Symbols DPP6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPP6. Recombinant protein control fragment (Product #RP-101264).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13483, 1804
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.