Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPP8 Antibody (OTI1D2), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271972R
Description
DPP8 Monoclonal specifically detects DPP8 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DPP8 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DPP8 | |
Full length recombinant protein of human DPP8 (NP_569118) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1D2 | |
Western Blot, Immunohistochemistry | |
dipeptidyl peptidase 8, Dipeptidyl peptidase IV-related protein 1, dipeptidyl peptidase IV-related protein-1, dipeptidylpeptidase 8, dipeptidyl-peptidase 8, DP8Dipeptidyl peptidase VIII, DPP VIII, DPRP-1, DPRP1Prolyl dipeptidase DPP8, EC 3.4.14.5, FLJ14920, FLJ20283, MGC26191, MSTP141 | |
Mouse | |
Affinity Purified | |
GPCR | |
54878.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction