Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP8 Monoclonal Antibody (OTI1D2)
GREENER_CHOICE

Catalog No. PIMA525895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525895 Supplier Invitrogen™ Supplier No. MA525895
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPP8 Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The encoded protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dpp8
Gene Accession No. Q6V1X1, Q80YA7
Gene Alias 2310004I03Rik; 4932434F09Rik; AI666706; dipeptidyl peptidase 8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; dipeptidyl peptidase VIII; dipeptidylpeptidase 8; dipeptidyl-peptidase 8; DP8; DPP VIII; DPP8; DPRP1; DPRP-1; MST097; MSTP097; MSTP135; MSTP141; Prolyl dipeptidase DPP8
Gene Symbols Dpp8
Host Species Mouse
Immunogen Full length human recombinant protein of DPP8 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 315758, 487605, 54878, 74388
Target Species Canine, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.