Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPP8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521646
Description
Recommended positive controls: A549, HeLa, NT2D1, mouse brain. Predicted reactivity: Mouse (93%), Rat (86%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The encoded protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
DPP8 | |
Polyclonal | |
Unconjugated | |
Dpp8 | |
2310004I03Rik; 4932434F09Rik; AI666706; dipeptidyl peptidase 8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; dipeptidyl peptidase VIII; dipeptidylpeptidase 8; dipeptidyl-peptidase 8; DP8; DPP VIII; DPP8; DPRP1; DPRP-1; MST097; MSTP097; MSTP135; MSTP141; Prolyl dipeptidase DPP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54878, 74388 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q6V1X1, Q80YA7 | |
Dpp8 | |
Recombinant fragment corresponding to a region within amino acids 349 and 662 of DPRP1 (Uniprot ID#Q6V1X1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction