Learn More
Invitrogen™ DPP8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532731
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Rabbit (100%); Dog, Bovine, Bat, Panda, Horse, Pig (94%); Marmoset, Elephant, Opossum (89%); Mouse, Rat (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This gene encodes a member of the peptidase S9B family, a small family of dipeptidyl peptidases that are able to cleave peptide substrates at a prolyl bond. The encoded protein shares similarity with dipeptidyl peptidase IV in that it is ubiquitously expressed, and hydrolyzes the same substrates. These similarities suggest that, like dipeptidyl peptidase IV, this protein may play a role in T-cell activation and immune function. Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
DPP8 | |
Polyclonal | |
Unconjugated | |
Dpp8 | |
2310004I03Rik; 4932434F09Rik; AI666706; dipeptidyl peptidase 8; Dipeptidyl peptidase IV-related protein 1; dipeptidyl peptidase IV-related protein-1; dipeptidyl peptidase VIII; dipeptidylpeptidase 8; dipeptidyl-peptidase 8; DP8; DPP VIII; DPP8; DPRP1; DPRP-1; MST097; MSTP097; MSTP135; MSTP141; Prolyl dipeptidase DPP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100346226, 54878 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.8 mg/mL | |
PBS with 0.1% sodium azide | |
Q6V1X1 | |
Dpp8 | |
Synthetic 18 amino acid peptide from internal region of human DPP8. | |
50 μg | |
Primary | |
Human, Rabbit, Monkey | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.