Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPP9 Monoclonal Antibody (OTI6A12), TrueMAB™, OriGene
SDP

Catalog No. 502540543
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0543 30 μL
50-167-4811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0543 Supplier OriGene Supplier No. TA504039S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPP9 Monoclonal antibody specifically detects DPP9 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DPP9
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6A12
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Enpep
Gene Accession No. P16406, Q07075
Gene Alias 6430584G11Rik; A330078I11; Dipeptidyl peptidase 9; dipeptidyl peptidase IV-related protein 2; dipeptidyl peptidase IV-related protein-2; dipeptidyl peptidase IX; Dipeptidyl peptidase-like protein 9; dipeptidylpeptidase 9; dipeptidyl-peptidase 9; DKFZp762F117; DP9; DPLP9; DPP IX; DPP9; DPRP2; DPRP-2; FLJ16073
Host Species Mouse
Immunogen Full length human recombinant protein of human ENPEP produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13809, 2028
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.