Learn More
DPP9 Mouse anti-Human, Clone: OTI2F10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503937

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein that is a member of the S9B family in clan SC of the serine proteases. The protein has been shown to have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Although the activity of this protein is similar to that of dipeptidyl peptidase 4 (DPP4), it does not appear to be membrane bound. In general, dipeptidyl peptidases appear to be involved in the regulation of the activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. Several transcript variants of this gene have been described but not fully characterized.Specifications
DPP9 | |
Monoclonal | |
Unconjugated | |
DPP9 | |
DP9, DPLP9, DPRP-2, DPRP2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
91039 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI2F10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86TI2 | |
DPP9 | |
Full length human recombit protein of human DPP9 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.