Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532734
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532734 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532734 Supplier Invitrogen™ Supplier No. PA532734
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gibbon, Marmoset, Mouse, Bovine, Dog, Horse (100%); Hamster, Panda (94%); Bat, Elephant, Turkey, Chicken (88%); Gorilla, Opossum (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene encodes a protein that is a member of the S9B family in clan SC of the serine proteases. The protein has been shown to have post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Although the activity of this protein is similar to that of dipeptidyl peptidase 4 (DPP4), it does not appear to be membrane bound. In general, dipeptidyl peptidases appear to be involved in the regulation of the activity of their substrates and have been linked to a variety of diseases including type 2 diabetes, obesity and cancer. Several transcript variants of this gene have been described but not fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPP9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DPP9
Gene Accession No. Q86TI2, Q8BVG4
Gene Alias 6430584G11Rik; A330078I11; Dipeptidyl peptidase 9; dipeptidyl peptidase IV-related protein 2; dipeptidyl peptidase IV-related protein-2; dipeptidyl peptidase IX; Dipeptidyl peptidase-like protein 9; dipeptidylpeptidase 9; dipeptidyl-peptidase 9; DKFZp762F117; DP9; DPLP9; DPP IX; DPP9; DPRP2; DPRP-2; FLJ16073
Gene Symbols DPP9
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human DPP9.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100062517, 224897, 485033, 521100, 91039
Target Species Human, Mouse, Canine, Monkey, Bovine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.