Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPA2 Antibody (Z31P3C4*F11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250230AF594
Description
DPPA2 Monoclonal specifically detects DPPA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
DPPA2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
DPPA2 | |
Peptide Sequence -KMMKRLMTVE | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, ELISA, Immunohistochemistry | |
Z31P3C4*F11 | |
Western Blot, ELISA, Immunohistochemistry | |
CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein | |
Mouse | |
Protein A purified | |
Developmental Biology, Stem Cells | |
151871 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction