Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPPA2 Monoclonal Antibody (OTI1G10)
GREENER_CHOICE

Catalog No. PIMA526429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526429 Supplier Invitrogen™ Supplier No. MA526429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPPA2 Monoclonal Antibody for Western Blot

The DPPA2 (Developmental Pluripotency Associated 2) gene is prominently expressed in embryonic stem cells and is critical for maintaining their pluripotency and self-renewal capabilities. Located on chromosome 3, DPPA2 belongs to a family of genes that play significant roles in early embryonic development and epigenetic remodeling. The expression of DPPA2 is tightly regulated through mechanisms such as DNA methylation, and it is involved in regulatory pathways that control the expression of core pluripotency factors. Beyond its role in stem cell biology, DPPA2 expression is also noted in various cancers, where it can become reactivated, suggesting its involvement in oncogenic processes. In the context of cancer, DPPA2, along with DPPA4, has been studied for its potential to facilitate cellular reprogramming and contribute to tumorigenesis. Given its expression pattern, DPPA2 serves as both a marker of pluripotent potential in stem cells and a potential target for cancer diagnostics and therapy due to its reactivation in malignant tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA2
Applications Western Blot
Classification Monoclonal
Clone OTI1G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DPPA2
Gene Accession No. Q7Z7J5
Gene Alias 2410088E07Rik; C80932; cancer/testis antigen 100; CT100; D19Mgi18; developmental pluripotency associated 2; developmental pluripotency-associated 2; developmental pluripotency-associated protein 2; DPPA2; ECAT15-2; ECSA; embryonic stem cell (ESC) associated transcript 15-2; embryonic stem cell-associated transcript 15-2 protein; LOC683282; PESCRG1; Phsecrg1; Pluripotent embryonic stem cell-related gene 1 protein; similar to developmental pluripotency-associated 2
Gene Symbols DPPA2, LOC683282
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-175 of DPPA2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151871, 607166, 683282
Target Species Canine, Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.