Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585868
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585868 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585868 Supplier Invitrogen™ Supplier No. PA585868
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPPA2 Polyclonal Antibody for Western Blot

The DPPA2 (Developmental Pluripotency Associated 2) gene is prominently expressed in embryonic stem cells and is critical for maintaining their pluripotency and self-renewal capabilities. Located on chromosome 3, DPPA2 belongs to a family of genes that play significant roles in early embryonic development and epigenetic remodeling. The expression of DPPA2 is tightly regulated through mechanisms such as DNA methylation, and it is involved in regulatory pathways that control the expression of core pluripotency factors. Beyond its role in stem cell biology, DPPA2 expression is also noted in various cancers, where it can become reactivated, suggesting its involvement in oncogenic processes. In the context of cancer, DPPA2, along with DPPA4, has been studied for its potential to facilitate cellular reprogramming and contribute to tumorigenesis. Given its expression pattern, DPPA2 serves as both a marker of pluripotent potential in stem cells and a potential target for cancer diagnostics and therapy due to its reactivation in malignant tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene DPPA2
Gene Accession No. Q9CWH0
Gene Alias 2410088E07Rik; C80932; cancer/testis antigen 100; CT100; D19Mgi18; developmental pluripotency associated 2; developmental pluripotency-associated 2; developmental pluripotency-associated protein 2; DPPA2; ECAT15-2; ECSA; embryonic stem cell (ESC) associated transcript 15-2; embryonic stem cell-associated transcript 15-2 protein; LOC683282; PESCRG1; Phsecrg1; Pluripotent embryonic stem cell-related gene 1 protein; similar to developmental pluripotency-associated 2
Gene Symbols DPPA2
Host Species Rabbit
Immunogen Synthetic peptide within mouse DPPA2 aa 1-50.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 73703
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.