Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPA4 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298786V
Description
DPPA4 Polyclonal specifically detects DPPA4 in Human samples. It is validated for Western Blot.Specifications
DPPA4 | |
Polyclonal | |
Western Blot | |
developmental pluripotency associated 4, developmental pluripotency-associated protein 4, FLJ10713,2410091M23Rik | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human DPPA4. | |
0.1 mL | |
Stem Cell Markers | |
55211 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction