Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPPA4 Monoclonal Antibody (OTI1C3)
GREENER_CHOICE

Catalog No. PIMA526356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526356 Supplier Invitrogen™ Supplier No. MA526356
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DPPA4 Monoclonal Antibody for Western Blot, ICC/IF

The DPPA4 gene encodes developmental pluripotency associated 4, a protein that plays a crucial role in maintaining stem cell pluripotency and facilitating early embryonic development. DPPA4 is primarily expressed in pluripotent stem cells, such as embryonic stem cells, where it contributes to the regulation of gene expression and chromatin structure necessary for pluripotency and differentiation. This protein is also involved in the establishment of epigenetic marks that influence developmental pathways, ensuring the proper formation of tissue types during embryogenesis. DPPA4 functions in stem cell renewal and differentiation, impacting cellular processes that are fundamental to early development. Beyond its roles in normal physiology, DPPA4 has been studied in the context of oncology, where its expression is often upregulated in certain cancers, indicating its potential as a biomarker for tumorigenic stem cell-like properties. Research continues to explore DPPA4's mechanisms and interactions to understand its contributions to development and its implications in disease states, particularly in cancer biology and regenerative medicine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA4
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DPPA4
Gene Accession No. Q7L190
Gene Alias 2410091M23Rik; C76608; developmental pluripotency associated 4; developmental pluripotency-associated protein 4; Dppa4; ECAT15-1; Embryonic stem cell-associated transcript 15-1 protein; LOC685378; similar to developmental pluripotency associated 4 isoform 1
Gene Symbols DPPA4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 9-254 of DPPA4 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55211
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.