Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPA4 Monoclonal Antibody (OTI2F6), TrueMAB™, OriGene
SDP

Catalog No. 501678024
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8024 Supplier OriGene Supplier No. TA800036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

May play a role in maintaining cell pluripotentiality.

Specifications

Antigen DPPA4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI2F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DPPA4
Gene Accession No. Q7L190
Gene Alias 2410091M23Rik
Gene Symbols DPPA4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 9-254 of human DPPA4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55211
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.