Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dppa4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983008
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3008 Supplier Bioss Supplier No. BS13595R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in maintaining cell pluripotentiality.

Specifications

Antigen Dppa4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DPPA4
Gene Accession No. Q7L190
Gene Alias 2410091M23Rik; C76608; developmental pluripotency associated 4; Developmental pluripotency-associated protein 4; Dppa4; ECAT15-1; Embryonic stem cell-associated transcript 15-1 protein; LOC685378; similar to developmental pluripotency associated 4 isoform 1
Gene Symbols DPPA4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Dppa4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55211
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.