Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPPA5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564174
Description
Immunogen sequence: MGTLPARRHI PPWVKVPEDL KDPEVFQVQT RLLKAIFGPD GSRIPYIEQV S Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
Involved in the maintenance of embryonic stem (ES) cell pluripotency. Dispensable for self-renewal of pluripotent ES cells and establishment of germ cells. Associates with specific target mRNAs.
Specifications
DPPA5 | |
Polyclonal | |
Unconjugated | |
DPPA5 | |
AA536857; developmental pluripotency associated 5; developmental pluripotency associated 5A; developmental pluripotency-associated 5 protein; Developmental pluripotency-associated protein 5A; Dppa5; Dppa5a; Ecat2; embryonal stem cell specific gene 1; embryonal stem cell-specific gene 1 protein; ES cell-associated transcript 2 protein; ESG1; ESG-1; ESG1ESG-1; hDPPA5; mDppa5; Ph34; Protein pH 34 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
340168 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NC42 | |
DPPA5 | |
Recombinant protein corresponding to Human DPPA5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction