Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPPA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547452
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547452 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547452 Supplier Invitrogen™ Supplier No. PA547452
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 40% cross-reactivity with recombinant mouse DPPA5 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Involved in the maintenance of embryonic stem (ES) cell pluripotency. Dispensable for self-renewal of pluripotent ES cells and establishment of germ cells. Associates with specific target mRNAs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPPA5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene DPPA5
Gene Accession No. A6NC42
Gene Alias AA536857; developmental pluripotency associated 5; developmental pluripotency associated 5A; developmental pluripotency-associated 5 protein; Developmental pluripotency-associated protein 5A; Dppa5; Dppa5a; Ecat2; embryonal stem cell specific gene 1; embryonal stem cell-specific gene 1 protein; ES cell-associated transcript 2 protein; ESG1; ESG-1; ESG1ESG-1; hDPPA5; mDppa5; Ph34; Protein pH 34
Gene Symbols DPPA5
Host Species Goat
Immunogen E. coli-derived recombinant human DPPA5/ESG1 Met1-Lys116.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340168
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.