Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPD Oxalate Solution For Sulfide Testing, Reagents

DPPD Oxalate Solution For Sulfide Testing, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CD0014004C

Catalog No. 01-335-438


Only null left
Add to Cart

Description

Description

Specifications

Specifications

DPPD Oxalate Solution For Sulfide Testing
7664-93-9 , 7732-18-5
N,N-Dimethyl-1,4-phenylenediamine Oxalate, 4-Ammoniophenyl(dimethyl)ammonium Oxalate
Glass Bottle
<1
Non-sterile
Tan
125°C
Reagent
1.4921
4 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.