Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPII/QPP/DPP7 Antibody (003), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289622APC
Description
DPPII/QPP/DPP7 Monoclonal antibody specifically detects DPPII/QPP/DPP7 in Human samples. It is validated for ELISASpecifications
DPPII/QPP/DPP7 | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
29952 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
003 | |
ELISA | |
carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human DPPII/QPP/DPP7 (Accession#: NP_037511.2; Met1-Leu492). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction