Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPII/QPP/DPP7 Antibody (035), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290774PCP
Description
DPPII/QPP/DPP7 Monoclonal antibody specifically detects DPPII/QPP/DPP7 in Mouse samples. It is validated for ELISASpecifications
DPPII/QPP/DPP7 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
29952 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
035 | |
ELISA | |
carboxytripeptidase, Dipeptidyl aminopeptidase II, dipeptidyl arylamidase II, dipeptidyl peptidase 2, Dipeptidyl peptidase 7, Dipeptidyl peptidase II, dipeptidylpeptidase 7, dipeptidyl-peptidase 7, dipeptidyl-peptidase II, DPP II, DPP2, DPPII, EC 3.4.14.2, QPP, Quiescent cell proline dipeptidase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse DPPII/QPP/DPP7 (Uniprot#: Q9ET221; Met1-Arg506). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction