Learn More
Description
- Designed for detection with a fluorescence microplate reader.
- Non-species-specific reactivity.
Specifications
Specifications
| Content And Storage | Storage of components varies. See protocol for specific instructions. |
| Detection Method | Fluorometric |
| Gene ID (Entrez) | 1803 |
| Gene Symbol | DPP4 |
| Product Line | Novus |
| Quantity | 96 Tests |
| Synonym | ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 |
| Type | Activity Assay Kit |
| Research Category | Adaptive Immunity, Cellular Markers, GPCR, Immunology |
| Species | Non-species specific |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
