Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPIV/CD26 Antibody (155202), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB9541Y
This item is not returnable.
View return policy
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow CytometrySpecifications
DPPIV/CD26 | |
Monoclonal | |
DyLight 680 | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
Mouse myeloma cell line NS0-derived recombinant mouse DPPIV/CD26, Extracellular domain | |
0.5 mL | |
Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
1803 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry | |
155202 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction