Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPIV/CD26 Antibody (236.3) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222434100UG
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
DPPIV/CD26 | |
Monoclonal | |
1 mg/ml | |
Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 1:10 - 1:500 | |
DPP4 | |
Rat 110-120 kD single chain glycoprotein Depeptidyl Peptidase IV (CD26). | |
100 ug | |
Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
1803 | |
Human, Rat | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
236.3 | |
Unconjugated | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody reacts with Rat. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction