Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPIV/CD26 Antibody (H207-1082) - BSA Free

Rat Monoclonal Antibody
$182.00 - $464.00
Specifications
Antigen | DPPIV/CD26 |
---|---|
Clone | H207-1082 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000 |
Applications | Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017528
![]() |
Novus Biologicals
NB100629970.025MG |
0.025 mg |
Each for $182.00
|
|
|||||
NB10062997
![]() |
Novus Biologicals
NB10062997 |
0.125 mg |
Each for $464.00
|
|
|||||
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
DPPIV/CD26 | |
1 mg/mL | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rat | |
Adaptive Immunity, GPCR, Immunology | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
DPP4 | |
IgG2a | |
Protein G purified | |
NB100-62997 recognises mouse CD26, which is also known as thymocyte-activating molecule (THAM). Murine CD26 is a type II integral membrane dipeptidyl-peptidase that is expressed at the cell surface as a heterodimer, of approximately 220 kDa, and has a soluble form in the plasma. The CD26 molecule is a multifunctional glycoprotein that has ectopeptidase activity and is also reported to be involved in cell adhesion, signal transduction in T lymphocytes and HIV infection. CD26 is differentially expressed on developing thymocytes and is upregulated on activated T-cells but is not expressed on resting and thioglycollate stimulated macrophages. Expression of CD26 has also been reported on cells from a variety of non-haematopoietic tissues including liver, intestine and kidney. |
H207-1082 | |
Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
1803 | |
Made to Mouse CD26 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title