Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPIV/CD26 Antibody (H207-1082), DyLight 350

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10062997UV
Description
DPPIV/CD26 Monoclonal specifically detects DPPIV/CD26 in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
DPPIV/CD26 | |
Monoclonal | |
DyLight 350 | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-62997 recognises mouse CD26, which is also known as thymocyte-activating molecule (THAM). Murine CD26 is a type II integral membrane dipeptidyl-peptidase that is expressed at the cell surface as a heterodimer, of approximately 220 kDa, and has a soluble form in the plasma. The CD26 molecule is a multifunctional glycoprotein that has ectopeptidase activity and is also reported to be involved in cell adhesion, signal transduction in T lymphocytes and HIV infection. CD26 is differentially expressed on developing thymocytes and is upregulated on activated T-cells but is not expressed on resting and thioglycollate stimulated macrophages. Expression of CD26 has also been reported on cells from a variety of non-haematopoietic tissues including liver, intestine and kidney. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry, ELISA | |
H207-1082 | |
Flow Cytometry, ELISA | |
DPP4 | |
Made to Mouse CD26 | |
0.1 mL | |
Adaptive Immunity, Cellular Markers, GPCR, Immunology | |
1803 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction