Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPPIV/CD26 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10061658
Description
DPPIV/CD26 Polyclonal specifically detects DPPIV/CD26 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
DPPIV/CD26 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Rat | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 ug/mL, Flow Cytometry 10 ug/mL, Immunohistochemistry 8 ug/mL, Immunohistochemistry-Paraffin 8 ug/mL, Peptide ELISA Detection limit 1:64000 | |
P28843, P14740 | |
DPP4 | |
Peptide with sequence C-PPHFDKSKKYP corresponding to internal region according to NP_001926.2. | |
0.1 mg | |
Adaptive Immunity, GPCR, Immunology | |
1803 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction