Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPPL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987155
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7155 Supplier Bioss Supplier No. BS6250R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Phosphatidate phosphatase (PAP) plays important role in lipid-signaling metabolism in eukaryotic cells. Two distinct types of PAP (PAP1 and PAP2) activity have been distinguished by their subcellular localization and differential sensitivity to N-ethylmaleimide (NEM) and Mg2+. A yeast diacylglycerol pyrophosphate (DGPP) phosphatase (DPP1) and mammalian DGPP phosphatase (PAP2) have been identified as Mg2+-independent and NEM-insensitive membrane-associated. PPAPDC1A (also known as DPPL2) and PPAPDC1B (DPPL1) form a novel type of Mg2+-independent and NEM-sensitive mammalian phosphatidate phosphatase showing broad substrate specificity. Knockdown experiments indicated that this protein is involved with multiple cell signaling pathways, including the JAK-Stat3, MAP kinase, and PKC pathways. PPAPDC1B may also potentiate the estrogen receptor pathway by down-regulating DUSP22.

Specifications

Antigen DPPL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLPP5
Gene Accession No. Q8NEB5
Gene Alias 1810019D05Rik; 2310022A04Rik; AW045471; diacylglycerol pyrophosphate like 1; diacylglycerol pyrophosphate phosphatase-like 1; DPPL1; HTPAP; phosphatidate phosphatase PPAPDC1B; phosphatidic acid phosphatase type 2 domain containing 1; phosphatidic acid phosphatase type 2 domain containing 1B; Phosphatidic acid phosphatase type 2 domain-containing protein 1B; Phospholipid phosphatase 5; PLPP5; Ppapdc1; PPAPDC1B; testicular secretory protein Li 38
Gene Symbols PLPP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DPPL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84513
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.