Learn More
Invitrogen™ DPT Polyclonal Antibody

Description
This antibody is predicted to react with bovine, mouse and porcine based on sequence homology.
Specifications
Specifications
| Antigen | DPT |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | DPT |
| Gene Accession No. | Q07507 |
| Gene Alias | 1810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:136613 |
| Gene Symbols | DPT |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.