Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528370 Supplier Invitrogen™ Supplier No. PA528370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Dermatopontin tansfected 293T cell. Predicted reactivity: Rat (93%), Xenopus laevis (81%), Dog (95%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPT
Applications Western Blot
Classification Polyclonal
Concentration 0.53 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene DPT
Gene Accession No. Q07507
Gene Alias 1810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:136613
Gene Symbols DPT
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Dermatopontin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1805
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.