Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569157
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569157 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569157 Supplier Thermo Scientific Supplier No. PA569157
Add to Cart
Add to Cart

DPT Polyclonal Antibody

DPT Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DPT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPT
Gene Accession No. Q9QZZ6
Gene Alias 1810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:136613
Gene Symbols DPT
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DPT (aa 45-94).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56429
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.