Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100064 Supplier Invitrogen™ Supplier No. PA5100064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Dermatopontin.

Dermatopontin is an extracellular matrix protein with possible functions in cell-matrix interactions and matrix assembly. The protein is found in various tissues and many of its tyrosine residues are sulphated. Dermatopontin is postulated to modify the behavior of TGF-beta through interaction with decorin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DPT
Gene Accession No. Q07507, Q9QZZ6
Gene Alias 1810032B19Rik; 22 kDa extracellular matrix protein; 5033416F05Rik; Dermatan sulfate proteoglycan-associated protein 22K; dermatopontin; Dpt; Early quiescence protein 1; early quiescence protein-1; Eq1; EQ-1; hypothetical protein LOC555282; si:dkey-98p3.2; similar to Dermatopontin precursor (Tyrosine-rich acidic matrix protein) (TRAMP); TRAMP; Tyrosine-rich acidic matrix protein; zgc:136613
Gene Symbols DPT
Host Species Rabbit
Immunogen A synthesized peptide derived from human DPT(Accession Q07507), corresponding to amino acid residues A58-N108.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1805, 289178, 56429
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.