Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPY19L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144055
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144055 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144055 Supplier Invitrogen™ Supplier No. PA5144055
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human Jurkat whole cell, human HepG2 whole cell. IHC: human rectum adenocarcinoma tissue, human ovarian serous adenocarcinoma tissue. Flow: HEL cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

Belongs to the DPY19 family. There are two named isoforms. The specific function of DPY19L1 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPY19L1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene DPY19L1
Gene Accession No. Q2PZI1
Gene Alias 1100001I19Rik; 6030402K12; AI225861; AW558481; dpy-19 like 1; dpy-19 like 1 (C. elegans); dpy-19 like C-mannosyltransferase 1; DPY19L1; dpy-19-like 1; dpy-19-like 1 (C. elegans); DPY-19-like protein 1; GA0500; KIAA0877; Probable C-mannosyltransferase DPY19L1; protein dpy-19 homolog 1; RGD1305822
Gene Symbols DPY19L1
Host Species Rabbit
Immunogen E.coli-derived human DPY19L1 recombinant protein (Position: L354-H657).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23333
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.