Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPY19L1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988437
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8437 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8437 Supplier Bioss Supplier No. BS8289R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Belongs to the DPY19 family. There are two named isoforms. The specific function of DPY19L1 is not yet known.

Specifications

Antigen DPY19L1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DPY19L1
Gene Accession No. Q2PZI1
Gene Alias 1100001I19Rik; 6030402K12; AI225861; AW558481; dpy-19 like 1; dpy-19 like 1 (C. elegans); dpy-19 like C-mannosyltransferase 1; DPY19L1; dpy-19-like 1; dpy-19-like 1 (C. elegans); Dpy-19-like protein 1; GA0500; KIAA0877; Probable C-mannosyltransferase DPY19L1; Protein dpy-19 homolog 1; RGD1305822
Gene Symbols DPY19L1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DPY19L1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23333
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.