Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPY19L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544405 Supplier Invitrogen™ Supplier No. PA544405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: IIGEFNNLPQ EELLQWIKYS TTSDAVFAGA MPTMASIKLS TLHPIVNHPH Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

Gene ontology (GO) annotation includes protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan; spermatid development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPY19L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPY19L2
Gene Accession No. Q6NUT2
Gene Alias 4932443J21Rik; dpy-19 like 2; Dpy19l2; dpy-19-like 2; dpy-19-like 2 (C. elegans); dpy-19-like protein 2; Dyp19l2; Gm18376; Probable C-mannosyltransferase DPY19L2; probable C-mannosyltransferase DPY19L2-like; protein dpy-19 homolog 2; RGD1564311; SPATA34; spermatogenesis associated 34; SPGF9; UNQ3127/PRO10284
Gene Symbols DPY19L2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DPY19L2.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283417
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.