Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPY19L2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$343.50 - $573.00

Specifications

Antigen DPY19L2
Dilution Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB169323
SDP
View Documents
Novus Biologicals
NBP31712325UL
25 μg
Each for $343.50
Only null left
Add to Cart
 
NB169322
SDP
View Documents
Novus Biologicals
NBP317123100UL
100 μg
Each for $573.00
Only null left
Add to Cart
 
Description

Description

DPY19L2 Polyclonal antibody specifically detects DPY19L2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

DPY19L2
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
dpy-19-like 2 (C. elegans), Dpy-19-like protein 2, FLJ32949, FLJ36166, protein dpy-19 homolog 2
This antibody was developed against Recombinant Protein corresponding to amino acids: KQGVSSKRLQSSGCSQSKGRRGASLAREPEVEEEM
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Polyclonal
Purified
RUO
PBS, pH 7.2, 40% glycerol
283417
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.