Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPY19L3 Polyclonal Antibody
DPY19L3 Polyclonal Antibody
Supplier: Thermo Scientific PA552545
Description
DPY19L3 Polyclonal Antibody for ICC/IF, IHC

Specifications
DPY19L3 | |
Polyclonal | |
Unconjugated | |
DPY19L3 | |
dpy-19 like 3 (C. elegans); DPY19L3; dpy-19-like 3; dpy-19-like protein 3; Probable C-mannosyltransferase DPY19L3; protein dpy-19 homolog 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
147991 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZPD9 | |
DPY19L3 | |
Recombinant protein corresponding to Human DPY19L3. Recombinant protein control fragment (Product #RP-89738). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction