Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPY19L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598526
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598526 Supplier Invitrogen™ Supplier No. PA598526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DPY19L3 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Probable C-mannosyltransferase that mediates C-mannosylation of tryptophan residues on target proteins. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPY19L3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DPY19L3
Gene Accession No. Q6ZPD9
Gene Alias dpy-19 like 3 (C. elegans); DPY19L3; dpy-19-like 3; dpy-19-like protein 3; Probable C-mannosyltransferase DPY19L3; protein dpy-19 homolog 3
Gene Symbols DPY19L3
Host Species Rabbit
Immunogen Recombinant Human Probable C-mannosyltransferase DPY19L3 protein (587-697AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147991
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.