Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPY19L4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528515
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable C-mannosyltransferase that mediates C-mannosylation of tryptophan residues on target proteins. [UniProt].
Specifications
DPY19L4 | |
Polyclonal | |
Unconjugated | |
DPY19L4 | |
dpy-19 like 4 (C. elegans); DPY19L4; dpy-19-like 4; dpy-19-like 4 (C. elegans); dpy-19-like protein 4; Gm1023; Narg3; NMDA receptor-regulated gene 3; probable C-mannosyltransferase DPY19L4; Protein dpy-19 homolog 4; RGD1305828 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
286148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q7Z388 | |
DPY19L4 | |
Recombinant fragment corresponding to a region within amino acids 506 and 723 of Human DPY19L4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction