Learn More
Invitrogen™ DPY30 Polyclonal Antibody

Description
Immunogen sequence: YGLTDNVERI VENEKINAEK SSKQKVDLQS LPTRAYLDQT VVPILLQGLA VLAKERPPNP IEFLASYLLK NKAQFEDRN Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Specifications
Specifications
| Antigen | DPY30 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | DPY30 |
| Gene Accession No. | Q9C005 |
| Gene Alias | 2810410M20Rik; Aip1; C87842; Cps25; dosage compensation-related protein DPY30; DPY30; dpy-30 histone methyltransferase complex regulatory subunit; dpy-30 homolog; dpy-30 homolog (C. elegans); dpy-30, histone methyltransferase complex regulatory subunit; Dpy-30L; dpy-30-like protein; HDPY-30; Protein dpy-30 homolog; protein rAIP1; Saf19; Unknown (protein for MGC:136993) |
| Gene Symbols | DPY30 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.