Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPY30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570474 Supplier Invitrogen™ Supplier No. PA570474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%.

As part of the MLL1/MLL complex, involved in the methylation of histone H3 at 'Lys-4', particularly trimethylation. Histone H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. May play some role in histone H3 acetylation. In a teratocarcinoma cell, plays a crucial role in retinoic acid-induced differentiation along the neural lineage, regulating gene induction and H3 'Lys-4' methylation at key developmental loci. May also play an indirect or direct role in endosomal transport.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPY30
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPY30
Gene Accession No. Q9C005
Gene Alias 2810410M20Rik; Aip1; C87842; Cps25; dosage compensation-related protein DPY30; DPY30; dpy-30 histone methyltransferase complex regulatory subunit; dpy-30 homolog; dpy-30 homolog (C. elegans); dpy-30, histone methyltransferase complex regulatory subunit; Dpy-30L; dpy-30-like protein; HDPY-30; Protein dpy-30 homolog; protein rAIP1; Saf19; Unknown (protein for MGC:136993)
Gene Symbols DPY30
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human DPY30.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84661
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.