Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPY30 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567360
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7360 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7360 Supplier Bethyl Laboratories, Inc Supplier No. A304296AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat and Bovine

As part of the MLL1/MLL complex, involved in the methylation of histone H3 at ′Lys-4′, particularly trimethylation. Histone H3 ′Lys-4′ methylation represents a specific tag for epigenetic transcriptional activation. May play some role in histone H3 acetylation. In a teratocarcinoma cell, plays a crucial role in retinoic acid-induced differentiation along the neural lineage, regulating gene induction and H3 ′Lys-4′ methylation at key developmental loci. May also play an indirect or direct role in endosomal transport.

Specifications

Antigen DPY30
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene DPY30
Gene Accession No. Q9C005
Gene Alias Cps25, dpy-30 histone methyltransferase complex regulatory subunit, dpy-30 homolog, dpy-30 homolog (C. elegans), dpy-30L, dpy-30-like protein, HDPY-30, Saf19
Gene Symbols DPY30
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84661
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.