Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DPY30 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00
Specifications
Antigen | DPY30 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
DPY30 Polyclonal specifically detects DPY30 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DPY30 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
dpy-30 homolog (C. elegans), dpy-30L, Dpy-30-like protein, HDPY-30, protein dpy-30 homolog, Saf19 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-99 of human DPY30 (NP_115963.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
modENCODE Antibodies | |
PBS with 50% glycerol, pH7.3. | |
84661 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title