Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPYD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109657 Supplier Invitrogen™ Supplier No. PA5109657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAPVLSKDSA DIESILALNP RTQTHATLCS TSAKKLDKKH WKRNPDKNCF NCEKLENNFD DIKHTTLGER GALREAMRCL KCADAPCQKS CPTNLDIKSF ITSIANKNYY GAAKMIFSDN PLGLTCGMVC PTSDLCVGGC NLYATEEGPI NIGGLQQFAT ETLILAFSLM NHL.

Involved in pyrimidine base degradation. Catalyzes the reduction of uracil and thymine. Also involved the degradation of the chemotherapeutic drug 5-fluorouracil.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPYD
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DPYD
Gene Accession No. Q12882, Q8CHR6
Gene Alias AI315208; DHP; DHPDHase; dihydropyrimidine dehydrogenase; dihydropyrimidine dehydrogenase [NADP(+)]; Dihydrothymine dehydrogenase; dihydrouracil dehydrogenase; DPD; Dpyd; E330028L06Rik
Gene Symbols DPYD
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-173 of human DPYD (NP_0001012).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1806, 99586
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.