Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DPYSL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543752
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543752 Supplier Invitrogen™ Supplier No. PA543752
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VFDLTTTPKG GTPAGSARGS PTRPNPPVRN LHQSGFSLSG TQVDEGVRSA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPYSL3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DPYSL3
Gene Accession No. Q14195
Gene Alias Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1
Gene Symbols DPYSL3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human DPYSL3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1809
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.