Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DPYSL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552760
Description
Immunogen sequence: IIDHVVPEPE SSLTEAYEKW REWADGKSCC DYALHVDITH WNDSVKQEVQ NLIKDKGVNS FMVYMAYKDL YQVSNTELYE IFTCLGELGA IAQVHAENGD IIAQEQTRML EMGITGPE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
DPYSL3 | |
Polyclonal | |
Unconjugated | |
DPYSL3 | |
Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.09 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14195 | |
DPYSL3 | |
Recombinant protein corresponding to Human DPYSL3. Recombinant protein control fragment (Product #RP-88673). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction