Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ DR 4485 Hydrochloride
GREENER_CHOICESDP

Catalog No. 500550RD Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-055-0RD 50 mg
50-051-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-055-0RD Supplier Tocris Bioscience™ Supplier No. 5005/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High affinity and selective 5-HT7 antagonist; orally bioavailable

DR 4485 hydrochloride is a high affinity and selective 5-HT7 antagonist (pKi = 8.14). Exhibits selectivity for 5-HT7 over other 5-HT receptors. Inhibits 5-HT-induced cAMP accumulation in HEK-293 cells expressing the 5-HT7 receptor. Orally bioavailable.

Specifications

Chemical Name or Material 6-Chloro-2a-[4-[4-(4-chlorophenyl)-3,6-dihydro-1(2H)-pyridinyl]butyl]-2a,3,4,5-tetrahydrobenz[cd]indol-2(1H)-one hydrochloride
CAS 402942-53-4
Quantity 50 mg
Molecular Formula C26H28Cl2N2O.HCl
Formula Weight 491.88
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.