Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dräger™ Carbon Monoxide (CO) XXS Replacement Sensor

Catalog No. 196810882 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured
19-681-0882 Carbon Monoxide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-681-0882 Supplier Dräger™ Supplier No. 6810882

XXS Replacement Sensor, for use with Pac 5500, Pac 7000, X-am 2000, X-am 5000 multi-gas detectors.

  • Measuring range: 0 to 2000 ppm
  • Resolution: 2 ppm
  • Warranty: 2 years
  • Carbon Monoxide (CO)

Specifications

Components Measured Carbon Monoxide
For Use With (Equipment) Pac 5500, Pac 7000, X-am 2000, X-am 5000 Multigas Detectors
Type Replacement Sensor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.