Learn More
Dräger™ Civil Defense Training Sets
For use with Civil Defense Simultest Set I and Set V
$123.37
Specifications
| For Use With (Equipment) | Civil Defense Simultest Training Set |
|---|
Alert:
These tubes are for Training ONLY.
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
19-822-154
|
Dräger™
8103230 |
Muriatic Acid (CDS Set I) |
Each for $123.37
|
|
|||||
|
19-822-155
|
Dräger™
8103240 |
Chlorine Tablets (CDS Set V) |
Each for $123.37
|
|
|||||
Description
- Cantain five tubes set mimics the tubes in CDS Set I and Set V
- Muriatic acid (CDS Set I) and Chlorine tablets (CDS Set V) can be used to show color change
Specifications
| Civil Defense Simultest Training Set |