Learn More
Dräger™ CMS Chips: Butadiene
Catalog No. 19640460 Shop All Draeger Safety, Inc. Products
Change view
Click to view available options
Measurement Range:
1 to 25
Catalog No. Measurement Range
19-640-460 1 to 25
1 options
Catalog No. 19-640-460 Supplier Dräger™ Supplier No. 6406460
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Butadiene CMS Chip
Humidity Range 5 to 30mg/L
Measurement Range 1 to 25
Response Time 90 to 550 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C